5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%

Lieferant: Apollo Scientific

PC908743-1G
APOSPC908743-1GEA 1770 EUR
APOSPC908743-1G
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid 97%
5-Chloro-4-fluoro-1H-indole-2-carboxylic acid
Formel: C₉H₅ClFNO₂
Molecular Weight: 213,593 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835295
CAS-Nummer: 186446-26-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR