5-Fluoro-2-hydroxypyrimidine 95%

Lieferant: Apollo Scientific

PC908749-1G
APOSPC908749-1GEA 46.4 EUR
APOSPC908749-1G
5-Fluoro-2-hydroxypyrimidine 95%
5-Fluoro-2-hydroxypyrimidine
Formel: C₄H₃FN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00223678
CAS-Nummer: 214290-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR