5-Fluoro-2-methyl-1H-indole-3-carbaldehyde 95%

Lieferant: Apollo Scientific

PC908750-1G PC908750-250MG
APOSPC908750-1GEA 318 EUR
APOSPC908750-1G APOSPC908750-250MG
5-Fluoro-2-methyl-1H-indole-3-carbaldehyde 95%
5-Fluoro-2-methyl-1H-indole-3-carbaldehyde
Formel: C₁₀H₈FNO
Molecular Weight: 177.175 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07643210
CAS-Nummer: 815586-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR