5-Fluoro-2-methyl-1H-indole-3-carbaldehyde 95%
Lieferant: Apollo Scientific
PC908750-1G
PC908750-250MG
APOSPC908750-1GEA
318
EUR
APOSPC908750-1G
APOSPC908750-250MG
5-Fluoro-2-methyl-1H-indole-3-carbaldehyde 95%
5-Fluoro-2-methyl-1H-indole-3-carbaldehyde
Formel:
C₁₀H₈FNO Molecular Weight: 177.175 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07643210 CAS-Nummer: 815586-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...