6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one 95%

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-3,4-dihydroisoquinolin-1(2H)-one

PC908805-1G
APOSPC908805-1GEA 60.9 EUR
APOSPC908805-1G
6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one 95%
6-Fluoro-3,4-dihydro-2H-isoquinolin-1-one
Formel: C₉H₈FNO
Molecular Weight: 165,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04114864
CAS-Nummer: 214045-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR