(3-Trifluoromethylpyrazol-1-yl)acetic acid 95%

Lieferant: Apollo Scientific

Synonyme: 2-(3-(Trifluoromethyl)-1H-pyrazol-1-yl)acetic acid

PC908826-500MG
APOSPC908826-500MGEA 538 EUR
APOSPC908826-500MG
(3-Trifluoromethylpyrazol-1-yl)acetic acid 95%
(3-Trifluoromethylpyrazol-1-yl)acetic acid
Formel: C₆H₅F₃N₂O₂
Molecular Weight: 194,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969231
CAS-Nummer: 926241-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR