2-(3-Fluorophenoxy)ethylamine hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Fluorophenoxy)ethylamine

PC908941-1G PC908941-5G
APOSPC908941-1GEA 190 EUR
APOSPC908941-1G APOSPC908941-5G
2-(3-Fluorophenoxy)ethylamine hydrochloride 98%
2-(3-Fluorophenoxy)ethylamine hydrochloride
Formel: C₈H₁₁ClFNO
Molecular Weight: 191.63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11099553
CAS-Nummer: 120351-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR