1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate 97%
Lieferant: Apollo Scientific
Synonyme:
Mal-C6-amine TFA
PC908951-250MG
APOSPC908951-250MGEA
102
EUR
APOSPC908951-250MG
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate 97%
1-(6-Aminohexyl)-1H-pyrrole-2,5-dione 2,2,2-trifluoroacetate
Formel:
C₁₀H₁₆N₂O₂·CF₃CO₂H Molecular Weight: 310,27 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519191 CAS-Nummer: 731862-92-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...