5-Fluor-2-mercaptobenzothiazol 98%
Lieferant: Apollo Scientific
Synonyme:
5-Fluorobenzo[d]thiazole-2-thiol
PC9090-1G
APOSPC9090-1GEA
108
EUR
APOSPC9090-1G
5-Fluor-2-mercaptobenzothiazol 98%
5-Fluoro-2-mercaptobenzothiazole
Formel:
C₇H₄FNS₂ Molecular Weight: 185,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181177 CAS-Nummer: 155559-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...