(2-Fluoro-benzoylamino)-acetic acid 98%
Lieferant: Apollo Scientific
PC909033-1G
APOSPC909033-1GEA
270
EUR
APOSPC909033-1G
(2-Fluoro-benzoylamino)-acetic acid 98%
(2-Fluoro-benzoylamino)-acetic acid
Formel:
C₉H₈FNO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00460020 CAS-Nummer: 363-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...