2-Methyl-4-(trifluoromethyl)benzaldehyde 95%

Lieferant: Apollo Scientific

PC909109-1G PC909109-5G
APOSPC909109-1GEA 119 EUR
APOSPC909109-1G APOSPC909109-5G
2-Methyl-4-(trifluoromethyl)benzaldehyde 95%
2-Methyl-4-(trifluoromethyl)benzaldehyde
Formel: C₉H₇F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18399853
CAS-Nummer: 888739-67-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR