3'-Fluor-4'-nitroacetanilid 98%

Lieferant: Apollo Scientific

Synonyme: N-(3-fluoro-4-nitrophenyl)acetamide

PC909137-5G
APOSPC909137-5GEA 249 EUR
APOSPC909137-5G
3'-Fluor-4'-nitroacetanilid 98%
3'-Fluor-4'-nitroacetanilid
Formel: C₈H₇FN₂O₃
Molecular Weight: 198.1512 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01463345
CAS-Nummer: 345-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR