6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%
Lieferant: Apollo Scientific
Synonyme:
6-Fluoro-4,4-dimethyl-3,4-dihydroquinolin-2(1H)-one
PC909146-25G
PC909146-5G
APOSPC909146-25GEA
857
EUR
APOSPC909146-25G
APOSPC909146-5G
6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%
6-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one
Formel:
C₁₁H₁₂FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD12913844 CAS-Nummer: 395673-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...