2-Fluoro-4-phenylphenol 96%

Lieferant: Apollo Scientific

PC909286-1G
APOSPC909286-1GEA 364 EUR
APOSPC909286-1G
2-Fluoro-4-phenylphenol 96%
2-Fluoro-4-phenylphenol
Formel: C₁₂H₉FO
Molecular Weight: 188,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14701828
CAS-Nummer: 84376-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR