3-Fluoro-4-phenylphenol 96%

Lieferant: Apollo Scientific

PC909287-1G
APOSPC909287-1GEA 328 EUR
APOSPC909287-1G
3-Fluoro-4-phenylphenol 96%
3-Fluoro-4-phenylphenol
Formel: C₁₂H₉FO
Molecular Weight: 188,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01814577
CAS-Nummer: 477860-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR