3-Fluoro-5-phenylphenol 95%
Lieferant: Apollo Scientific
PC909288-1G
APOSPC909288-1GEA
364
EUR
APOSPC909288-1G
3-Fluoro-5-phenylphenol 95%
3-Fluoro-5-phenylphenol
Formel:
C₁₂H₉FO Molecular Weight: 188,198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14701829 CAS-Nummer: 187392-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...