3-Fluoro-5-phenylbenzonitrile 96%

Lieferant: Apollo Scientific

PC909663-1G
APOSPC909663-1GEA 364 EUR
APOSPC909663-1G
3-Fluoro-5-phenylbenzonitrile 96%
3-Fluoro-5-phenylbenzonitrile
Formel: C₁₃H₈FN
Molecular Weight: 197,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14701726
CAS-Nummer: 1214352-57-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR