Methyl (2,3-difluorophenyl)acetate 96%
Lieferant: Apollo Scientific
PC909686-1G
APOSPC909686-1GEA
435
EUR
APOSPC909686-1G
Methyl (2,3-difluorophenyl)acetate 96%
Methyl (2,3-difluorophenyl)acetate
Formel:
C₉H₈F₂O₂ Molecular Weight: 186,155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025022 CAS-Nummer: 1036273-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...