Methyl (E)-3-(3,5-difluorophenyl)prop-2-enoate 95%

Lieferant: Apollo Scientific

PC909694-1G
APOSPC909694-1GEA 435 EUR
APOSPC909694-1G
Methyl (E)-3-(3,5-difluorophenyl)prop-2-enoate 95%
Methyl (E)-3-(3,5-difluorophenyl)prop-2-enoate
Formel: C₁₀H₈F₂O₂
Molecular Weight: 198,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124795
CAS-Nummer: 705250-75-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR