Methyl 3-fluoro-5-methylbenzoate 98%
Lieferant: Apollo Scientific
PC909705-1G
APOSPC909705-1GEA
178
EUR
APOSPC909705-1G
Methyl 3-fluoro-5-methylbenzoate 98%
Methyl 3-fluoro-5-methylbenzoate
Formel:
C₉H₉FO₂ Molecular Weight: 168,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204167 CAS-Nummer: 660416-38-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...