2,5-Difluoro-4-methylbenzoic acid methyl ester 95%
Lieferant: Apollo Scientific
PC909715-1G
APOSPC909715-1GEA
632
EUR
APOSPC909715-1G
2,5-Difluoro-4-methylbenzoic acid methyl ester 95%
2,5-Difluoro-4-methylbenzoic acid methyl ester
Formel:
C₉H₈F₂O₂ Molecular Weight: 186.155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16250137 CAS-Nummer: 1355488-71-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...