Methyl 2-(3-bromo-4-fluorophenyl)acetate 95%

Lieferant: Apollo Scientific

PC909726-1G
APOSPC909726-1GEA 434 EUR
APOSPC909726-1G
Methyl 2-(3-bromo-4-fluorophenyl)acetate 95%
Methyl 2-(3-bromo-4-fluorophenyl)acetate
Formel: C₉H₈BrFO₂
Molecular Weight: 247,061 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025009
CAS-Nummer: 885681-61-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR