N-(Trifluoromethylthio)phthalimide

Lieferant: Apollo Scientific
Achtung

PC909739-1G PC909739-5G
APOSPC909739-1GEA 232 EUR
APOSPC909739-1G APOSPC909739-5G
N-(Trifluoromethylthio)phthalimide
N-(Trifluoromethylthio)phthalimide
Formel: C₉H₄F₃NO₂S
Schmelzpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18449627
CAS-Nummer: 719-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR