Methyl 2-fluoro-4-methylbenzoate 95%

Lieferant: Apollo Scientific

PC909749-25G
APOSPC909749-25GEA 1220 EUR
APOSPC909749-25G
Methyl 2-fluoro-4-methylbenzoate 95%
Methyl 2-fluoro-4-methylbenzoate
Formel: C₉H₉FO₂
Molecular Weight: 168,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204168
CAS-Nummer: 74733-29-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR