Methyl 3-fluoro-2-methylbenzoate 95%

Lieferant: Apollo Scientific

PC909755-1G
APOSPC909755-1GEA 71.2 EUR
APOSPC909755-1G
Methyl 3-fluoro-2-methylbenzoate 95%
Methyl 3-fluoro-2-methylbenzoate
Formel: C₉H₉FO₂
Molecular Weight: 168,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06203865
CAS-Nummer: 230301-81-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR