Methyl 3-methyl-5-(trifluoromethyl)benzoate

Lieferant: Apollo Scientific

PC909774-1G
APOSPC909774-1GEA 661 EUR
APOSPC909774-1G
Methyl 3-methyl-5-(trifluoromethyl)benzoate
Methyl 3-methyl-5-(trifluoromethyl)benzoate
Formel: C₁₀H₉F₃O₂
Molecular Weight: 218,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698072
CAS-Nummer: 161622-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR