Methyl 3,5-difluoroisonicotinate 95%

Lieferant: Apollo Scientific

PC909781-1G
APOSPC909781-1GEA 376 EUR
APOSPC909781-1G
Methyl 3,5-difluoroisonicotinate 95%
Methyl 3,5-difluoroisonicotinate
Formel: C₇H₅F₂NO₂
Molecular Weight: 173.117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698040
CAS-Nummer: 1214324-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR