Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%

Lieferant: Apollo Scientific
Achtung

PC909782-1G
APOSPC909782-1GEA 29 EUR
APOSPC909782-1G
Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%
Methyl-2-chloro-5-(trifluoromethyl)nicotinate
Formel: C₈H₅ClF₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD22559950
CAS-Nummer: 1360934-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR