Methyl 2,6-difluoro-4-hydroxybenzoate 98%

Lieferant: Apollo Scientific

PC909789-1G
APOSPC909789-1GEA 248 EUR
APOSPC909789-1G
Methyl 2,6-difluoro-4-hydroxybenzoate 98%
Methyl 2,6-difluoro-4-hydroxybenzoate
Formel: C₈H₆F₂O₃
Molecular Weight: 188,128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042796
CAS-Nummer: 194938-88-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR