4,6-Difluoro-1,3-benzoxazol-2-amine 96%
Lieferant: Apollo Scientific
PC909840-1G
APOSPC909840-1GEA
828
EUR
APOSPC909840-1G
4,6-Difluoro-1,3-benzoxazol-2-amine 96%
4,6-Difluoro-1,3-benzoxazol-2-amine
Formel:
C₇H₄F₂N₂O Molecular Weight: 170,116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD17216199 CAS-Nummer: 1339452-03-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...