2-Ethoxy-1-fluoro-3-nitrobenzene 95%

Lieferant: Apollo Scientific

PC909881-1G
APOSPC909881-1GEA 0 EUR
APOSPC909881-1G
2-Ethoxy-1-fluoro-3-nitrobenzene 95%
2-Ethoxy-1-fluoro-3-nitrobenzene
Formel: C₈H₈FNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD17014041
CAS-Nummer: 1233954-79-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR