4-Ethoxy-3-fluoronitrobenzene 95%

Lieferant: Apollo Scientific

PC909886-1G
APOSPC909886-1GEA 538 EUR
APOSPC909886-1G
4-Ethoxy-3-fluoronitrobenzene 95%
4-Ethoxy-3-fluoronitrobenzene
Formel: C₈H₈FNO₃
Molecular Weight: 185,152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12547857
CAS-Nummer: 326-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR