1-Fluoro-2-nitro-3-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

PC909896-1G
APOSPC909896-1GEA 719 EUR
APOSPC909896-1G
1-Fluoro-2-nitro-3-(propan-2-yloxy)benzene 95%
1-Fluoro-2-nitro-3-(propan-2-yloxy)benzene
Formel: C₉H₁₀FNO₃
Molecular Weight: 199,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13689267
CAS-Nummer: 1233951-63-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR