1,2-Difluoro-4-methoxy-5-nitrobenzene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC909902-1G
PC909902-250MG
APOSPC909902-1GEA
140
EUR
APOSPC909902-1G
APOSPC909902-250MG
1,2-Difluoro-4-methoxy-5-nitrobenzene 95%
1,2-Difluoro-4-methoxy-5-nitrobenzene
Formel:
C₇H₅F₂NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11040445 CAS-Nummer: 66684-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...