4-Bromo-2,3-difluoropyridine 95%

Lieferant: Apollo Scientific

PC909991-1G
APOSPC909991-1GEA 177 EUR
APOSPC909991-1G
4-Bromo-2,3-difluoropyridine 95%
4-Bromo-2,3-difluoropyridine
Formel: C₅H₂BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD16609910
CAS-Nummer: 1227597-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR