2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide 97%

Lieferant: Apollo Scientific

PC9100-10G PC9100-5G
APOSPC9100-10GEA 105 EUR
APOSPC9100-10G APOSPC9100-5G
2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide 97%
2-Chloro-N-[4-(trifluoromethyl)phenyl]acetamide
Formel: C₉H₇ClF₃NO
Schmelzpunkt: 158…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084933
CAS-Nummer: 2707-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR