3-Chloro-6-fluoro-2-methylpyridine 98%
Lieferant: Apollo Scientific
PC910084-1G
APOSPC910084-1GEA
538
EUR
APOSPC910084-1G
3-Chloro-6-fluoro-2-methylpyridine 98%
3-Chloro-6-fluoro-2-methylpyridine
Formel:
C₆H₅ClFN Molecular Weight: 145,562 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185447 CAS-Nummer: 1214342-64-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...