3-Chloro-2-fluoro-6-methylpyridine 95%

Lieferant: Apollo Scientific

PC910085-1G
APOSPC910085-1GEA 729 EUR
APOSPC910085-1G
3-Chloro-2-fluoro-6-methylpyridine 95%
3-Chloro-2-fluoro-6-methylpyridine
Formel: C₆H₅ClFN
Molecular Weight: 145,562 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16610389
CAS-Nummer: 1227574-35-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR