4-Chloro-2,3-difluorophenol 95%

Lieferant: Apollo Scientific

PC910092-1G
APOSPC910092-1GEA 538 EUR
APOSPC910092-1G
4-Chloro-2,3-difluorophenol 95%
4-Chloro-2,3-difluorophenol
Formel: C₆H₃ClF₂O
Molecular Weight: 164,537 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415633
CAS-Nummer: 1261634-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR