1-Chloro-2,3-difluoro-4-methoxybenzene 95%

Lieferant: Apollo Scientific

PC910093-1G
APOSPC910093-1GEA 632 EUR
APOSPC910093-1G
1-Chloro-2,3-difluoro-4-methoxybenzene 95%
1-Chloro-2,3-difluoro-4-methoxybenzene
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415548
CAS-Nummer: 1261808-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR