2,4-Difluoro-3-chloroanisole 95%
Lieferant: Apollo Scientific
PC910097-1G
APOSPC910097-1GEA
0
EUR
APOSPC910097-1G
2,4-Difluoro-3-chloroanisole 95%
2,4-Difluoro-3-chloroanisole
Formel:
C₇H₅ClF₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD18415544 CAS-Nummer: 1261472-07-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...