1-Chloro-3,5-difluoro-2-methoxybenzene 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-4,6-difluoroanisole

PC910098-1G
APOSPC910098-1GEA 632 EUR
APOSPC910098-1G
1-Chloro-3,5-difluoro-2-methoxybenzene 95%
1-Chloro-3,5-difluoro-2-methoxybenzene
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415543
CAS-Nummer: 943830-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR