4-Chlor-2,5-difluoranisol 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-2,5-difluoro-4-methoxybenzene

PC910099-1G
APOSPC910099-1GEA 632 EUR
APOSPC910099-1G
4-Chlor-2,5-difluoranisol 95%
4-Chlor-2,5-difluoranisol
Formel: C₇H₅ClF₂O
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1261846-22-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR