2-Chloro-1-fluoro-3-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

PC910110-1G
APOSPC910110-1GEA 632 EUR
APOSPC910110-1G
2-Chloro-1-fluoro-3-(propan-2-yloxy)benzene 95%
2-Chloro-1-fluoro-3-(propan-2-yloxy)benzene
Formel: C₉H₁₀ClFO
Molecular Weight: 188,627 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23114120
CAS-Nummer: 1369889-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR