5-Chloro-2-fluoro-3-methylbenzaldehyde 95%

Lieferant: Apollo Scientific

PC910112-1G
APOSPC910112-1GEA 632 EUR
APOSPC910112-1G
5-Chloro-2-fluoro-3-methylbenzaldehyde 95%
5-Chloro-2-fluoro-3-methylbenzaldehyde
Formel: C₈H₆ClFO
Molecular Weight: 172,584 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26687656
CAS-Nummer: 1538090-76-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR