4-Chlor-2,3-difluorbenzaldehyd 95%
Lieferant: Apollo Scientific
PC910118-1G
APOSPC910118-1GEA
538
EUR
APOSPC910118-1G
4-Chlor-2,3-difluorbenzaldehyd 95%
4-Chloro-2,3-difluorobenzaldehyde
Formel:
C₇H₃ClF₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD11845976 CAS-Nummer: 1160573-23-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...