2-Chloro-3,4-difluoroanisole 95%

Lieferant: Apollo Scientific

PC910119-1G
APOSPC910119-1GEA 632 EUR
APOSPC910119-1G
2-Chloro-3,4-difluoroanisole 95%
2-Chloro-3,4-difluoroanisole
Formel: C₇H₅ClF₂O
Molecular Weight: 178,564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415541
CAS-Nummer: 1261769-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR