1-Chloro-4,5-difluoro-2-methoxybenzene 95%

Lieferant: Apollo Scientific

PC910121-1G
APOSPC910121-1GEA 632 EUR
APOSPC910121-1G
1-Chloro-4,5-difluoro-2-methoxybenzene 95%
1-Chloro-4,5-difluoro-2-methoxybenzene
Formel: C₇H₅ClF₂O
Molecular Weight: 178.564 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18415542
CAS-Nummer: 1261598-22-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR