1-Chloro-2,4-difluoro-5-methoxybenzene 95%

Lieferant: Apollo Scientific

PC910122-1G
APOSPC910122-1GEA 0 EUR
APOSPC910122-1G
1-Chloro-2,4-difluoro-5-methoxybenzene 95%
1-Chloro-2,4-difluoro-5-methoxybenzene
Formel: C₇H₅ClF₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD18415547
CAS-Nummer: 1261628-93-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR