1-Chloro-3-fluoro-5-(propan-2-yloxy)benzene 95%
Lieferant: Apollo Scientific
PC910126-1G
APOSPC910126-1GEA
632
EUR
APOSPC910126-1G
1-Chloro-3-fluoro-5-(propan-2-yloxy)benzene 95%
1-Chloro-3-fluoro-5-(propan-2-yloxy)benzene
Formel:
C₉H₁₀ClFO Molecular Weight: 188,627 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23114122 CAS-Nummer: 1369947-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...